Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010229.2 | NP_034359.2 | 3 | 444 | ||
GenBank mRNA | AK045865.1 | - | 3 | 267 | ||
AK149292.1 | - | 3 | 262 | |||
AK163640.1 | - | 3 | 445 | |||
BC109003.1 | - | 3 | 237 | |||
BC109004.1 | - | 3 | 249 | |||
M64689.1 | - | 3 | 250 | |||
X59398.1 | - | 3 | 301 |