Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008052.3 | NP_032078.2 | 5 | 2006 | ||
GenBank mRNA | AB015422.1 | - | 4 | 1609 | ||
AK088630.1 | - | 5 | 1962 | |||
AK169455.1 | - | 5 | 2007 | |||
AK220514.1 | - | 4 | 1587 | |||
BC053055.1 | - | 5 | 1152 | 5685525 | ||
BC065075.1 | - | 6 | 1788 | |||
U38252.1 | - | 3 | 1191 |