Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113188.1 | NP_001106659.1 | 14 | 1552 | ||
NM_001113189.1 | NP_001106660.1 | 15 | 1465 | |||
NM_008053.2 | NP_032079.1 | 15 | 1465 | |||
GenBank mRNA | AF124385.1 | - | 10 | 966 | ||
AK049231.1 | - | 6 | 515 | |||
BC019139.1 | - | 15 | 1446 | 5041635 | ||
CB196254.1 | - | 2 | 88 | |||
CF170707.1 | - | 2 | 130 | |||
X90875.1 | - | 15 | 1444 |