Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301298.1 | NP_001288227.1 | 5 | 2127 | ||
NM_021356.2 | NP_067331.2 | 5 | 2087 | |||
GenBank mRNA | AJ250669.1 | - | 5 | 1234 | ||
AK132176.1 | - | 5 | 2128 | |||
AK155423.1 | - | 5 | 2098 | |||
BC007483.1 | - | 5 | 2087 | 3495926 | ||
BC094659.1 | - | 5 | 1438 | 6854020 | ||
CT010264.1 | - | 5 | 1211 |