Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008067.4 | NP_032093.3 | 7 | 1036 | ||
GenBank mRNA | AK039144.1 | - | 7 | 977 | ||
AK134760.1 | - | 7 | 945 | |||
BC119317.1 | - | 7 | 834 | 8734066 | ||
BC120686.1 | - | 7 | 834 | 40129609 | ||
M86568.1 | - | 7 | 938 |