Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133661.3 | NP_598422.2 | 14 | 1874 | ||
GenBank mRNA | BC019211.1 | - | 14 | 1878 | 5053116 | |
M97632.1 | - | 14 | 1614 |