Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160404.1 | NP_001153876.1 | 9 | 1327 | ||
NM_013814.3 | NP_038842.3 | 9 | 1415 | |||
GenBank mRNA | AK083391.1 | - | 9 | 1331 | ||
AK148550.1 | - | 3 | 395 | |||
BC056215.1 | - | 9 | 1332 | 30019829 | ||
BC090962.1 | - | 8 | 1205 | 30626890 | ||
U73820.1 | - | 7 | 1080 |