Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015737.4 | NP_056552.1 | 1 | 1036 | ||
GenBank mRNA | AK033494.1 | - | 1 | 1001 | ||
AK148036.1 | - | 1 | 1004 | |||
AK153253.1 | - | 1 | 1014 | |||
BC057882.1 | - | 1 | 1027 | 5350623 | ||
U73819.1 | - | 1 | 756 |