Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008108.4 | NP_032134.2 | 1 | 315 | ||
GenBank mRNA | AK082877.1 | - | 1 | 236 | ||
AK146174.1 | - | 1 | 241 | |||
AK152457.1 | - | 1 | 316 | |||
AK160533.1 | - | 1 | 273 | |||
AK167962.1 | - | 1 | 241 | |||
BC101963.1 | - | 1 | 230 | 40045099 | ||
BC101964.1 | - | 1 | 230 | 40045100 | ||
BC101965.1 | - | 1 | 230 | |||
BC103565.1 | - | 1 | 229 | 40045101 | ||
L06443.1 | - | 1 | 331 | |||
S52658.1 | - | 1 | 271 |