Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gdf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008110.2 | NP_032136.2 | 2 | 534 | ||
GenBank mRNA | AK136145.1 | - | 2 | 474 | ||
BC052667.1 | - | 2 | 464 | 30054181 | ||
L06444.1 | - | 2 | 442 |