Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131020.1 | NP_001124492.1 | 4, 5 | 842 | ||
NM_010277.3 | NP_034407.2 | 4, 5 | 842 | |||
GenBank mRNA | AF332061.1 | - | 4, 5 | 669 | ||
AF332062.1 | - | 4, 5 | 669 | |||
AK079577.1 | - | 4, 5 | 776 | |||
AK140151.1 | - | 4, 5 | 774 | |||
BC100728.1 | - | 4, 5 | 721 | |||
BC100737.1 | - | 4, 5 | 714 | |||
BC101968.1 | - | 4, 5 | 721 | |||
BC103571.1 | - | 4, 5 | 715 | |||
BC139357.1 | - | 4, 5 | 842 | 8862379 | ||
BC139358.1 | - | 4, 5 | 842 | 8862380 | ||
CX219900.1 | - | 2, 3 | 186 | |||
EF101554.1 | - | 4, 5 | 488 | |||
K01347.1 | - | 4, 5 | 678 | |||
M25937.1 | - | 4, 5 | 678 |