Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008175.4 | NP_032201.2 | 12 | 1708 | ||
GenBank mRNA | AK013678.1 | - | 12 | 1524 | ||
AK018744.1 | - | 12 | 1549 | |||
AK147932.1 | - | 12 | 1525 | |||
AK149570.1 | - | 12 | 1526 | |||
AK150205.1 | - | 12 | 1524 | |||
AK150338.1 | - | 12 | 1513 | |||
AK150420.1 | - | 12 | 1512 | |||
AK150717.1 | - | 12 | 1524 | |||
AK150746.1 | - | 12 | 1526 | |||
AK150881.1 | - | 12 | 1526 | |||
AK151007.1 | - | 12 | 1524 | |||
AK151162.1 | - | 12 | 1526 | |||
AK151343.1 | - | 12 | 1524 | |||
AK151713.1 | - | 12 | 1524 | |||
AK151756.1 | - | 12 | 1524 | |||
AK152048.1 | - | 12 | 1524 | |||
AK152087.1 | - | 12 | 1524 | |||
AK152118.1 | - | 12 | 1513 | |||
AK152213.1 | - | 12 | 1513 | |||
AK152705.1 | - | 12 | 1524 | |||
AK152765.1 | - | 12 | 1524 | |||
AK152944.1 | - | 12 | 1513 | |||
AK153183.1 | - | 12 | 1514 | |||
AK153269.1 | - | 12 | 1523 | |||
AK153289.1 | - | 12 | 1524 | |||
AK153418.1 | - | 12 | 1524 | |||
AK153470.1 | - | 12 | 1524 | |||
AK154492.1 | - | 12 | 1524 | |||
AK159397.1 | - | 12 | 1513 | |||
AK159428.1 | - | 12 | 1513 | |||
AK159557.1 | - | 12 | 1524 | |||
AK159809.1 | - | 12 | 1524 | |||
AK160022.1 | - | 12 | 1513 | |||
AK160033.1 | - | 12 | 1524 | |||
AK160074.1 | - | 12 | 1513 | |||
AK185467.1 | - | 1 | 37 | |||
BC037047.1 | - | 11 | 1505 | |||
BC049943.1 | - | 12 | 1510 | |||
M86736.1 | - | 12 | 1506 | |||
X62321.1 | - | 11 | 1489 |