Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294138.1 | NP_001281067.1 | 7 | 936 | ||
NM_001294140.1 | NP_001281069.1 | 6 | 852 | |||
NM_008194.3 | NP_032220.1 | 6 | 852 | |||
NM_212444.2 | NP_997609.1 | 6 | 852 | |||
GenBank mRNA | AK044308.1 | - | 6 | 690 | ||
AK088373.1 | - | 6 | 671 | |||
AK169424.1 | - | 7 | 742 | |||
AK169525.1 | - | 6 | 853 | |||
AK201906.1 | - | 1 | 57 | |||
BC003767.1 | - | 6 | 655 | 3593896 | ||
CB950214.1 | - | 1 | 34 | |||
U48403.1 | - | 6 | 629 |