- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Gene Symbol: Gyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294138.1 | NP_001281067.1 | 18, 19 | 1890 | ||
NM_001294140.1 | NP_001281069.1 | 18, 19 | 1824 | |||
NM_008194.3 | NP_032220.1 | 17, 18 | 1806 | |||
NM_212444.2 | NP_997609.1 | 17, 18 | 1806 | |||
GenBank mRNA | AK044308.1 | - | 18, 19 | 1662 | ||
AK088373.1 | - | 17, 18 | 1625 | |||
AK169424.1 | - | 18, 19 | 1696 | |||
AK169525.1 | - | 17, 18 | 1807 | |||
BC003767.1 | - | 17, 18 | 1609 | 3593896 | ||
CF740682.1 | - | 2, 3 | 285 | |||
U48403.1 | - | 17, 18 | 1583 |