Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030678.3 | NP_109603.2 | 7 | 1237 | ||
GenBank mRNA | AK050813.1 | - | 5 | 670 | ||
AK085476.1 | - | 3 | 263 | |||
AK169344.1 | - | 7 | 1233 | |||
AK170404.1 | - | 7 | 1236 | |||
AK170585.1 | - | 7 | 1157 | |||
AK171148.1 | - | 7 | 1192 | |||
AK171426.1 | - | 7 | 1216 | |||
BC019389.1 | - | 7 | 1030 | 3663506 | ||
BC131687.1 | - | 7 | 1084 | 40111722 | ||
BC131688.1 | - | 7 | 1084 | 40111723 | ||
BC152550.1 | - | 7 | 1084 | 40111724 | ||
U53218.1 | - | 7 | 1075 | |||
X94616.1 | - | 7 | 1084 |