Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008204.2 | NP_032230.1 | 2 | 151 | ||
GenBank mRNA | AY302212.1 | - | 2 | 151 | ||
AY302213.1 | - | 2 | 151 | |||
AY302214.1 | - | 2 | 151 | |||
AY302215.1 | - | 2 | 151 | |||
AY302216.1 | - | 2 | 151 | |||
AY302217.1 | - | 2 | 151 | |||
BC144938.1 | - | 2 | 174 | |||
BC150676.1 | - | 2 | 174 | 9087587 |