Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010404.3 | NP_034534.1 | 9 | 1204 | ||
NM_177981.2 | NP_817090.1 | 9 | 1204 | |||
GenBank mRNA | AJ000262.1 | - | 9 | 1150 | ||
AJ002271.1 | - | 9 | 1150 | |||
AK138436.1 | - | 1153 | ||||
BC034089.1 | - | 9 | 1142 | 4481735 | ||
BC053043.1 | - | 9 | 1191 | 5694628 |