Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hells | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008234.3 | NP_032260.2 | 10 | 1034 | ||
GenBank mRNA | AF155210.1 | - | 10 | 1034 | ||
AK013266.1 | - | 10 | 1073 | |||
AK147126.1 | - | 10 | 1065 | |||
AK166636.1 | - | 10 | 1034 | |||
BC090671.1 | - | 10 | 1079 | |||
BC100394.1 | - | 10 | 1031 | 30480539 | ||
U25691.2 | - | 10 | 1067 |