Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010424.4 | NP_034554.2 | 5 | 1087 | ||
GenBank mRNA | AK009581.1 | - | 5 | 1088 | ||
AK088986.1 | - | 5 | 1083 | |||
AK150697.1 | - | 5 | 1105 |