Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289458.1 | NP_001276387.1 | 4 | 555 | ||
NM_001289459.1 | NP_001276388.1 | 4 | 544 | |||
NM_001289460.1 | NP_001276389.1 | 4 | 555 | |||
NM_001289461.1 | NP_001276390.1 | 3 | 377 | |||
NM_010427.5 | NP_034557.3 | 4 | 527 | |||
GenBank mRNA | AF042856.1 | - | 3 | 356 | ||
AK042121.1 | - | 4 | 473 | |||
AK052289.1 | - | 4 | 556 | |||
AK082461.1 | - | 3 | 527 | |||
AK142159.1 | - | 4 | 473 | |||
BC119228.1 | - | 4 | 414 | 8733977 | ||
BC144764.1 | - | 4 | 426 | |||
D10212.1 | - | 3 | 350 | |||
D10213.1 | - | 3 | 350 | |||
S71816.1 | - | 3 | 350 | |||
X72307.1 | - | 3 | 411 | |||
X84046.1 | - | 3 | 390 |