Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hmmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013552.2 | NP_038580.2 | 4 | 316 | ||
GenBank mRNA | AF031932.1 | - | 4 | 301 | ||
AF079222.1 | - | 4, 5 | 265 | |||
AK020144.1 | - | 4 | 271 | |||
AK041617.1 | - | 4 | 296 | |||
AK143123.1 | - | 4 | 316 | |||
AK163410.1 | - | 4 | 242 | |||
AK164410.1 | - | 4 | 290 | |||
AK164597.1 | - | 3, 4 | 293 | |||
BC021427.1 | - | 4 | 314 | 5035341 | ||
CN455388.1 | - | 4 | 320 |