Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010465.2 | NP_034595.2 | 1 | 61 | ||
GenBank mRNA | AB221685.1 | - | 1 | 17 | ||
BC028482.1 | - | 1 | 61 | |||
BC141061.1 | - | 1 | 40 | 9055673 | ||
BX528590.1 | - | 1 | 62 | |||
X16511.1 | - | 1 | 144 |