Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hoxd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013554.5 | NP_038582.2 | 1 | 108 | ||
GenBank mRNA | AK134330.1 | - | 1 | 109 | ||
BC013463.1 | - | 1 | 69 | 4240579 | ||
BC048690.1 | - | 1 | 51 | 6516538 |