Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172812.2 | NP_766400.1 | 3 | 2402 | ||
GenBank mRNA | AK053140.1 | - | 3 | 2402 | ||
BC108972.1 | - | 3 | 1431 | 40047362 | ||
BC108973.1 | - | 3 | 1431 | 40047364 | ||
S49542.1 | - | 3 | 1309 |
Set Membership: |
![]() |