Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008311.2 | NP_032337.2 | 2 | 506 | ||
GenBank mRNA | AF498254.1 | - | 1 | 179 | ||
AF498255.1 | - | 1 | 179 | |||
AK033713.1 | - | 2 | 506 | |||
BC023690.1 | - | 2 | 455 | 5344328 | ||
Z15119.1 | - | 1 | 214 |
Set Membership: |
![]() |