Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010511.2 | NP_034641.1 | 6 | 951 | ||
GenBank mRNA | AF128216.1 | - | 6 | 902 | ||
AF128217.1 | - | 6 | 902 | |||
AK087525.1 | - | 6 | 940 | |||
AK150503.1 | - | 6 | 900 | |||
AK151079.1 | - | 6 | 905 | |||
AK152085.1 | - | 6 | 900 | |||
AK152384.1 | - | 6 | 900 | |||
AK153084.1 | - | 6 | 900 | |||
AK153347.1 | - | 6 | 903 | |||
AK159359.1 | - | 6 | 905 | |||
AK159660.1 | - | 6 | 845 | |||
AK159839.1 | - | 6 | 845 | |||
AK169593.1 | - | 6 | 920 | |||
AY157836.1 | - | 6 | 902 | |||
AY157837.1 | - | 6 | 902 | |||
BC132599.1 | - | 6 | 852 | 40130876 | ||
BC138617.1 | - | 6 | 852 | 8861637 | ||
J05265.1 | - | 6 | 928 | |||
M25764.1 | - | 6 | 853 | |||
M26711.1 | - | 6 | 928 | |||
M28233.1 | - | 6 | 924 | |||
M28995.1 | - | 6 | 829 |