Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igtp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018738.4 | NP_061208.3 | 3 | 676 | ||
GenBank mRNA | AK002843.1 | - | 3 | 614 | ||
AK088315.1 | - | 3 | 675 | |||
AK149817.1 | - | 3 | 605 | |||
AK151779.1 | - | 3 | 616 | |||
AK151860.1 | - | 3 | 597 | |||
AK152483.1 | - | 3 | 597 | |||
AK153436.1 | - | 3 | 616 | |||
AK169307.1 | - | 3 | 635 | |||
AY225483.1 | - | 2 | 241 | |||
BC132557.1 | - | 2 | 304 | 40130834 | ||
BC138607.1 | - | 2 | 304 | 8861627 | ||
U53219.1 | - | 3 | 588 |