Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Il18r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161842.1 | NP_001155314.1 | 8 | 1223 | ||
NM_001161843.1 | NP_001155315.1 | 8 | 1095 | |||
NM_008365.2 | NP_032391.1 | 7 | 1148 | |||
GenBank mRNA | AK087129.1 | - | 7 | 1098 | ||
AK089237.1 | - | 8 | 1224 | |||
BC020296.1 | - | 7 | 910 | |||
BC023240.1 | - | 8 | 1064 | 4483261 | ||
U43673.1 | - | 7 | 1148 |