Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008398.2 | NP_032424.2 | 7 | 1280 | ||
GenBank mRNA | AK157612.1 | - | 7 | 1287 | ||
BM950653.1 | - | 3 | 438 | |||
L23423.1 | - | 7 | 1278 |