Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008405.3 | NP_032431.2 | 7 | 767 | ||
GenBank mRNA | AF051367.1 | - | 7 | 752 | ||
AK036640.1 | - | 7 | 767 | |||
AK137534.1 | - | 7 | 768 | |||
AK158108.1 | - | 7 | 768 | |||
BC113142.1 | - | 7 | 736 | 40047633 |