Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145884.1 | NP_001139356.1 | 10 | 1678 | ||
NM_010580.2 | NP_034710.2 | 10 | 1678 | |||
GenBank mRNA | AF022110.1 | - | 10 | 1677 | ||
AF043256.1 | - | 10 | 1583 | |||
AF043257.1 | - | 10 | 1583 | |||
AK087476.1 | - | 10 | 1425 | |||
AK140251.1 | - | 10 | 1590 | |||
AK143532.1 | - | 10 | 1613 | |||
BC029170.1 | - | 5 | 525 | |||
BC046280.1 | - | 10 | 1587 | 6394641 | ||
BC058246.1 | - | 10 | 1642 | 6313668 |