Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008408.4 | NP_032434.3 | 3, 4 | 213 | ||
GenBank mRNA | AK030363.1 | - | 3, 4 | 277 | ||
AK077877.1 | - | 3, 4 | 237 | |||
AK081547.1 | - | 3, 4 | 269 | |||
AK087470.1 | - | 3, 4 | 252 | |||
AK129027.1 | - | 3, 4 | 251 | |||
AK150640.1 | - | 3, 4 | 258 | |||
AK151541.1 | - | 3, 4 | 269 | |||
AK151791.1 | - | 3, 4 | 269 | |||
AK152242.1 | - | 3, 4 | 258 | |||
AK153840.1 | - | 3, 4 | 214 | |||
AK153886.1 | - | 3, 4 | 314 | |||
AK160037.1 | - | 3, 4 | 252 | |||
AK160607.1 | - | 3, 4 | 214 | |||
AK168505.1 | - | 3, 4 | 258 | |||
AK168937.1 | - | 3, 4 | 258 | |||
AK169669.1 | - | 3, 4 | 237 | |||
AK189214.1 | - | 3, 4 | 237 | |||
AX799090.1 | - | 3, 4 | 256 | |||
BC028897.1 | - | 2, 3 | 164 | |||
BC037612.1 | - | 3, 4 | 252 | 5353797 | ||
BC085313.1 | - | 3, 4 | 242 | 30443880 | ||
L34260.1 | - | 3, 4 | 256 |