Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039056.2 | NP_001034145.1 | 4 | 1081 | ||
NM_001039057.2 | NP_001034146.1 | 4 | 1173 | |||
NM_001271687.1 | NP_001258616.1 | 5 | 1245 | |||
NM_001271689.1 | NP_001258618.1 | 5 | 1212 | |||
NM_001271690.1 | NP_001258619.1 | 5 | 1173 | |||
NM_001271691.1 | NP_001258620.1 | 4 | 1153 | |||
NM_001271692.1 | NP_001258621.1 | 4 | 1141 | |||
NM_001271693.1 | NP_001258622.1 | 4 | 1108 | |||
NM_001271694.1 | NP_001258623.1 | 4 | 1096 | |||
NM_001271695.1 | NP_001258624.1 | 3 | 1055 | |||
NM_019664.5 | NP_062638.1 | 5 | 1257 | |||
GenBank mRNA | AJ012368.1 | - | 4 | 1039 | ||
AJ300831.1 | - | 1 | 705 | |||
AK085689.1 | - | 3 | 915 | |||
AK143004.1 | - | 4 | 962 | |||
AK163000.1 | - | 3 | 1131 | |||
AK165346.1 | - | 5 | 1081 | |||
AK165436.1 | - | 4 | 1082 | |||
AY377988.1 | - | 2 | 728 | |||
BC010794.1 | - | 4 | 947 | 4221516 | ||
BC057915.1 | - | 5 | 1072 | 5042647 | ||
BC138477.1 | - | 3 | 954 | 8861497 | ||
BC138478.1 | - | 3 | 966 | 8861498 | ||
BC145288.1 | - | 3 | 954 | 9053842 |