Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039000.4 | NP_001034089.1 | 19 | 2625 | ||
NM_008447.4 | NP_032473.2 | 19 | 2625 | |||
GenBank mRNA | AF067179.1 | - | 19 | 2332 | ||
AK147352.1 | - | 19 | 2402 | |||
AK147660.1 | - | 19 | 2334 | |||
AK147707.1 | - | 19 | 2356 | |||
AK220479.1 | - | 19 | 2610 | |||
BC058396.1 | - | 19 | 2342 | 6824963 |