Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133957.1 | - | 12 | 1348 | ||
AK205309.1 | - | 1 | 43 | |||
BC040362.1 | - | 12 | 1409 | 4506177 | ||
BC049100.1 | - | 12 | 1607 | 5698182 | ||
D50366.1 | - | 12 | 1522 | |||
D50367.1 | - | 12 | 1522 |