Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017959.2 | NP_001017959.1 | 1 | 177 | ||
NM_001290485.1 | NP_001277414.1 | 1 | 177 | |||
NM_010685.4 | NP_034815.2 | 1 | 177 | |||
GenBank mRNA | AK012149.1 | - | 1 | 136 | ||
AK032974.1 | - | 1 | 139 | |||
AK048174.1 | - | 1 | 119 | |||
AK078193.1 | - | 1 | 171 | |||
AK088804.1 | - | 1 | 154 | |||
AK136550.1 | - | 1 | 138 | |||
AK139871.1 | - | 1 | 132 | |||
AK146731.1 | - | 1 | 158 | |||
AK150766.1 | - | 1 | 146 | |||
AK159272.1 | - | 1 | 132 | |||
AK159731.1 | - | 1 | 132 | |||
AK163933.1 | - | 1 | 178 | |||
BC138718.1 | - | 1 | 64 | 8861740 | ||
BC138723.1 | - | 1 | 64 | 8861745 | ||
BY283193.1 | - | 1 | 172 | |||
J05287.1 | - | 1 | 104 |