Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198911.1 | NP_001185840.1 | 9 | 1216 | ||
NM_001198912.1 | NP_001185841.1 | 8 | 1084 | |||
NM_001198913.1 | NP_001185842.1 | 8 | 1078 | |||
NM_008487.3 | NP_032513.3 | 8 | 1094 | |||
GenBank mRNA | AB093254.1 | - | 8 | 1167 | ||
AF177032.1 | - | 8 | 1061 | |||
AK035683.1 | - | 2 | 943 | |||
AK144972.1 | - | 9 | 1197 | |||
AK156379.1 | - | 8 | 1075 | |||
AK158232.1 | - | 9 | 1287 | |||
AK167628.1 | - | 8 | 1048 | |||
AK171223.1 | - | 8 | 1037 | |||
BC006589.1 | - | 8 | 1015 | 3587800 | ||
KC508090.1 | - | 8 | 1070 | |||
U28495.1 | - | 8 | 1061 | |||
X95761.1 | - | 8 | 952 |