Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026666.3 | NP_080942.1 | 16 | 3209 | ||
GenBank mRNA | AK084046.1 | - | 2 | 571 | ||
AK137727.1 | - | 4 | 675 | |||
AK147571.1 | - | 16 | 3210 | |||
BC060723.1 | - | 14 | 2137 | |||
BC098372.1 | - | 14 | 2369 | 30363389 |