Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Idh3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130884.4 | NP_570954.1 | 5, 6 | 421 | ||
GenBank mRNA | AY035212.1 | - | 5, 6 | 402 | ||
BC009022.1 | - | 5, 6 | 410 | 3257667 | ||
DN176034.1 | - | 5, 6 | 387 |