Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205343.1 | NP_001192272.1 | 27 | 3712 | ||
NM_130887.3 | NP_570957.2 | 26 | 3646 | |||
GenBank mRNA | AF314171.1 | - | 26 | 3619 | ||
AK053198.1 | - | 27 | 3714 | |||
AK086767.1 | - | 26 | 3629 | |||
BC005747.1 | - | 8 | 1183 | |||
BC132475.1 | - | 26 | 3586 | 40130752 | ||
BC137854.1 | - | 26 | 3586 | 8860873 | ||
BC144975.1 | - | 27 | 3652 | 9053523 |