Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 18 | 2126 | ||
NM_001252486.1 | NP_001239415.1 | 18 | 2126 | |||
NM_133247.3 | NP_573510.2 | 18 | 2150 | |||
GenBank mRNA | AF383174.1 | - | 18 | 2139 | ||
AK082251.1 | - | 18 | 2109 | |||
AK082943.1 | - | 18 | 2109 | |||
AK087467.1 | - | 18 | 2127 | |||
AK122442.1 | - | 15 | 4608 | |||
AK143844.1 | - | 18 | 2118 | |||
BC005506.1 | - | 18 | 2079 | |||
BC031366.1 | - | 10 | 1266 | |||
BC036980.1 | - | 11 | 1326 | |||
BC049870.1 | - | 5 | 391 | |||
BC067399.1 | - | 17 | 1992 | |||
BC089315.1 | - | 18 | 2072 |