Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172439.3 | NP_766027.2 | 3 | 1385 | ||
GenBank mRNA | AK034272.1 | - | 3 | 1386 | ||
AK136908.1 | - | 3 | 1372 | |||
AK165718.1 | - | 3 | 1756 | |||
AK170114.1 | - | 3 | 1351 | |||
AK171029.1 | - | 3 | 1346 | |||
BC022915.1 | - | 3 | 1338 | |||
BC046561.1 | - | 2 | 198 | |||
BC051452.1 | - | 3 | 1355 | |||
BC131634.1 | - | 3 | 1380 | 40096940 | ||
BC131635.1 | - | 3 | 1377 | 40096943 |