Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010771.6 | NP_034901.2 | 4 | 1304 | ||
GenBank mRNA | AK029510.1 | - | 3 | 682 | ||
AK087939.1 | - | 4 | 1189 | |||
AK088351.1 | - | 4 | 1197 | |||
AK129199.1 | - | 4 | 1255 | |||
AK135937.1 | - | 3 | 1037 | |||
AK157744.1 | - | 3 | 400 | |||
AK161128.1 | - | 4 | 1203 | |||
AK189430.1 | - | 3 | 226 | |||
BC029070.1 | - | 4 | 1215 | 3989914 | ||
BC090833.1 | - | 3 | 1077 | 6854095 | ||
CT010322.1 | - | 3 | 988 |