Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010771.6 | NP_034901.2 | 10, 11 | 2059 | ||
GenBank mRNA | AK029510.1 | - | 9, 10 | 1437 | ||
AK087939.1 | - | 10, 11 | 1944 | |||
AK088351.1 | - | 10, 11 | 1952 | |||
AK129199.1 | - | 10, 11 | 2010 | |||
AK135937.1 | - | 9, 10 | 1792 | |||
AK148384.1 | - | 3, 4 | 386 | |||
AK157744.1 | - | 9, 10 | 1155 | |||
AK161128.1 | - | 10, 11 | 1958 | |||
BC029070.1 | - | 10, 11 | 1970 | 3989914 | ||
BC090833.1 | - | 9, 10 | 1832 | 6854095 | ||
CT010322.1 | - | 9, 10 | 1743 |