Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008563.2 | NP_032589.1 | 3 | 289 | ||
GenBank mRNA | AK088142.1 | - | 3 | 338 | ||
AK133850.1 | - | 3 | 354 | |||
AK145563.1 | - | 3 | 388 | |||
AK146497.1 | - | 3 | 302 | |||
AK147065.1 | - | 3 | 304 | |||
AK148006.1 | - | 3 | 334 | |||
BC020400.1 | - | 3 | 304 | |||
BC031700.1 | - | 3 | 289 | 5007886 | ||
X62154.1 | - | 2 | 233 |