Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142169.1 | - | 2 | 158 | ||
AK165778.1 | - | 2 | 159 | |||
BC052854.1 | - | 2 | 160 | 30058638 | ||
BY100700.1 | - | 2 | 186 | |||
Z12604.1 | - | 2 | 146 |