Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008610.2 | NP_032636.1 | 10, 11 | 1919 | ||
GenBank mRNA | AK148184.1 | - | 10, 11 | 1918 | ||
AK163248.1 | - | 2, 3 | 231 | |||
AK179499.1 | - | 1, 2 | 121 | |||
AK189970.1 | - | 1, 2 | 119 | |||
AK190237.1 | - | 1, 2 | 119 | |||
AK190797.1 | - | 1, 2 | 119 | |||
AK190805.1 | - | 1, 2 | 119 | |||
AK190966.1 | - | 1, 2 | 119 | |||
AK191078.1 | - | 1, 2 | 121 | |||
AK191758.1 | - | 1, 2 | 119 | |||
AK192637.1 | - | 1, 2 | 119 | |||
AK192709.1 | - | 1, 2 | 119 | |||
AK193625.1 | - | 1, 2 | 119 | |||
AK193954.1 | - | 1, 2 | 119 | |||
AK195150.1 | - | 1, 2 | 119 | |||
AK195198.1 | - | 1, 2 | 119 | |||
AK195892.1 | - | 1, 2 | 120 | |||
AK197081.1 | - | 207 | ||||
AK197653.1 | - | 1, 2 | 119 | |||
AK198474.1 | - | 1, 2 | 119 | |||
AK198537.1 | - | 1, 2 | 119 | |||
AK198639.1 | - | 1, 2 | 119 | |||
AK198965.1 | - | 1, 2 | 120 | |||
AK199060.1 | - | 1, 2 | 119 | |||
AK199919.1 | - | 146 | ||||
AK200339.1 | - | 1, 2 | 119 | |||
AK200683.1 | - | 1, 2 | 119 | |||
AK201563.1 | - | 1, 2 | 119 | |||
AK201782.1 | - | 1, 2 | 119 | |||
AK202079.1 | - | 1, 2 | 119 | |||
AK202191.1 | - | 1, 2 | 120 | |||
AK203427.1 | - | 1, 2 | 119 | |||
AK203661.1 | - | 1, 2 | 119 | |||
AK203919.1 | - | 1, 2 | 119 | |||
AK204187.1 | - | 1, 2 | 120 | |||
AK204502.1 | - | 1, 2 | 119 | |||
AK204539.1 | - | 1, 2 | 119 | |||
AK204698.1 | - | 1, 2 | 119 | |||
AK204944.1 | - | 1, 2 | 119 | |||
AK219401.1 | - | 1, 2 | 119 | |||
BC070430.1 | - | 10, 11 | 1919 | 6813184 | ||
M84324.1 | - | 10, 11 | 1665 |