Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008628.2 | NP_032654.1 | 7 | 1153 | ||
GenBank mRNA | AK051960.1 | - | 7 | 1150 | ||
AK157842.1 | - | 7 | 1166 | |||
AK166179.1 | - | 7 | 1139 | |||
AK206893.1 | - | 2 | 69 | |||
BC047117.1 | - | 7 | 1161 | 4951173 | ||
BC050897.1 | - | 7 | 1136 | 6401835 | ||
U21011.1 | - | 7 | 1170 | |||
X81143.1 | - | 7 | 1153 |