Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Map4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205330.1 | NP_001192259.1 | 11, 12 | 2606 | ||
NM_001205332.1 | NP_001192261.1 | 11, 12 | 2606 | |||
NM_008633.4 | NP_032659.2 | 11, 12 | 2606 | |||
GenBank mRNA | AK004694.2 | - | 3, 4 | 878 | ||
AK087930.1 | - | 3, 4 | 891 | |||
AK146790.1 | - | 4, 5 | 1985 | |||
AK166558.1 | - | 2, 3 | 305 | |||
BC042645.1 | - | 4, 5 | 1979 | 5357023 | ||
BC044654.1 | - | 11, 12 | 2516 | 5400526 | ||
BC050893.1 | - | 11, 12 | 2481 | 5706356 | ||
BC055332.1 | - | 11, 12 | 2487 | 6407218 | ||
BC055364.1 | - | 10, 11 | 2446 | 30057231 | ||
M72414.1 | - | 11, 12 | 2910 |