Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008636.4 | NP_032662.3 | 7 | 1176 | ||
GenBank mRNA | AK030284.1 | - | 7 | 1155 | ||
AY260849.1 | - | 6 | 1030 | |||
AY260850.1 | - | 6 | 1030 | |||
BC017679.1 | - | 7 | 1138 | 4210829 | ||
X71327.1 | - | 7 | 1126 |